Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Supelpure™ HC Hydrocarbon Trap

The best trap available for removing hydrocarbons and other organics from carrier, fuel, air, and other gas streams

$225.00 - $607.00

Specifications

Product Line Supelpure™
Type Hydrocarbon Trap
For Use With (Application) Removing Hydrocarbons and Other Organics From Carrier, Fuel, Air, and Other Gas Streams
Includes Carbon Molecular Sieve
View More Specs

Products 4
Catalog Number Mfr. No. Capacity (Metric) Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Size Price Quantity & Availability  
11-102-0821
View Documents
MilliporeSigma™ Supelco™
22445U
120 cc 1/8 in.
Each for $240.50
Only null left
Add to Cart
 
11-101-0329
View Documents
MilliporeSigma™ Supelco™
22446
120 cc 1/4 in.
Each for $225.00
Only null left
Add to Cart
 
11-100-9057
View Documents
MilliporeSigma™ Supelco™
24518
750 cc 1/4 in.
Each for $607.00
Only null left
Add to Cart
 
11-100-9058
View Documents
MilliporeSigma™ Supelco™
24519
750 cc 1/2 in.
Each for $555.50
Only null left
Add to Cart
 
Description

Description

  • Primarily used for trapping hydrocarbons
  • Used in GC for carrier gas purification
  • Contains a carbon molecular sieve that also restricts the elution of moisture and oxygen
  • Surges of moisture and oxygen that enter the gas line during the installation of a new gas cylinder are released slowly, and diluted by the clean carrier gas from the new cylinder
  • Activated charcoal adsorbs hydrocarbons and other contaminants from carrier gases, air, and hydrogen
  • Particularly removes C3 and other higher hydrocarbons
  • Operates efficiently for approximately six months (when total hydrocarbons in the incoming gas average 10 ppm)
Specifications

Specifications

Supelpure™
Hydrocarbon Trap
Removing Hydrocarbons and Other Organics From Carrier, Fuel, Air, and Other Gas Streams
Carbon Molecular Sieve
Videos
SDS
Documents

Documents

Product Certifications

Supelpure is a registered trademark of Sigma-Aldrich Co. LLC

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.