Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-2A Packed Column Ferrule, 1/4 in.
Catalog No. 111009296 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
10/Pk.
50/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
11-100-9296 50/Pk.
11-101-0963 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-9296 Supplier MilliporeSigma™ Supelco™ Supplier No. 22471
Add to Cart
Add to Cart
Characteristics: Seals at 1/4 turn past fingertight. High reusability. Won't stick to metal or glass. Material Composition: DuPont VESPEL SP-21 (85% polyimide, 15% graphite). Maximum Temperature: 400°C
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 1/4 in.
Diameter (English) Inner 1/4 in.
Packaging Quantity 50/Pk.
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC