Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-4 Packed Column Ferrule, 1/4 in.
Catalog No. 111009527 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
10/Pk.
50/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
11-100-9527 50/Pk.
11-101-1212 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-9527 Supplier MilliporeSigma™ Supelco™ Supplier No. 22478
Add to Cart
Add to Cart
Supelco has refined the design of these graphite ferrules to provide a clean, sharp profile with minimal flash.
Characteristics: Seals at one-quarter turn past fingertight. Maximum sealing surface contact, reduced risk of column contamination at installation .Material Composition: flexible graphite. Maximum Temperature: 450°C
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 1/4 in.
Diameter (English) Inner 1/4 in.
Packaging Quantity 50/Pk.
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC