Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Super Concentrate Cleaner
Shop All Brady Corporation Products
Click to view available options
Quantity:
12 Ea.
4 Pk.
Description
- Heavy-Duty Cleaning Power: Specifically formulated to cut through stubborn dirt, grime and old adhesive residue.
- Ensures Optimal Adhesion: Maximizes floor tape bond and durability by ensuring even the most challenging surfaces are clean.
- Green and Eco-Friendly: Safe, non-hazardous formula.
- Versatile Application: Effectively cleans a wide range of floor types.
- Just Add to Water: Single-use packet saves storage space and allows for quick mixing.
Specifications
Specifications
| Product Type | Cleaner Concentrate |
| Color-coded | No |
| Form | Concentrated Liquid |
| Scent | Original |
| Sterility | Non-sterile |
| Quantity | 4 Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction