Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Super Magnet Lab Kit

Click to view available options
Grade:
1 to 6
Description
Includes
- 100 magnetic chips
- 100 magnetic paper clips
- 10 marbles
- Seven magnetic wands
- Two metal ring magnets
- Two mini metal horseshoe magnets
- Two metal bar magnets
- 5 in. plastic horseshoe magnet
- Activity guide and durable storage box
Everyone gets hands-on exploration with this complete 224-piece set.
Order Info
Grades 1 to 6.
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School |
| Grade | 1 to 6 |
| Product Type | Super Magnet Lab Kit |
| Includes | 100 Magnetic chips, 100 magnetic paper clips, 10 marbles, 7 magnetic wands, 2 Metal ring magnets, 2 mini Metal horseshoe magnets, 1 to5 in. Plastic horseshoe magnet, 2 Metal bar magnets activity guide and durable storage box |
| Age | 6 to 11 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction