Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Super-Q™ Plus High-Volume Water Purification System
GSA_VA

Catalog No. p-170497 Shop All MilliporeSigma Products
Click to view available options
Type:
With Pump

Catalog No. ZFSQ115P4

Supplier: MilliporeSigma™ ZFSQ115P4

Only null left
Add to Cart
Add to Cart
Request a Quote

Requires

Required filter cartridges each sold separately

  • Ion-Ex™ cartridge removes inorganics (CPMB 022 02)
  • Organex-Q™ cartridge removes trace organic contaminants (CPEX 02201)
  • For applications requiring particulate prefiltration and/or ultralow bacteria levels, contact your Fisher Scientific Customer Service Representative

With a Millipore Super-Q™ Plus High-Volume Water Purification System, you can easily produce up to 12L/min. of Type I ultrapure water

High-flow water systems provide final polish to water which has been pretreated by reverse osmosis, deionization or distillation. Eliminates need for water storage systems. Consistenlty produce ultrapure quality water that exceeds Type I ASTM™/CAP/CLSi specifications.

 

  • Water in bowls is automatically recirculated hourly to maintain water quality between uses
  • Gauge indicates pressure of incoming feedwater Display shows resistivity measurement Modular design allows customization for specific applications Also available with 1/3hp pump motor to move gravity-fed feedwater at a controlled flowrate and equalize flowrate in pressurized loops

 

Applications:

  • Produce water ideal for preparing tissue culture media and reagents, and as rinse water for electronics.

Order Info

Systems shown here are 4-bowl systems; 3-bowl system without pump (ZFSQ 115 03) is available through your Fisher Scientific Customer Service Representative.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Contaminant Rejection (Microbiological) Less than10cfu/mL
Daily Output >20 L
Flow Rate 10 to 12
Product Water Type Type 1
Resistivity 18.2
Total Organic Carbon <20
Type With Pump
Model Super-Q Plus (4-bowl)
Dimensions (L x W x H) 8.75 x 45.25 x 31.5 in. (22 x 94.5 x 80 cm)
Length (English) 8.75 in.
Length (Metric) 22 cm
Width (English) 45.25 in.
Width (Metric) 94.5 cm
Height (English) 31.5 in.
Height (Metric) 80 cm
Depth (English) 8.7 in.
Depth (Metric) 22 cm
Weight (English) 140 lb.
Weight (Metric) 63.5 kg
Voltage 120 V
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.