Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tians Super Track Boot Covers

Catalog No. p-4014957 Shop All Tians International Products
Click to view available options
Size:
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

For Cleanroom use

  • Disposable
  • Virgin polyethylene laminated to polypropylene
  • With non-woven conductive/dissipative strip
  • High top
  • Height: 7 in.
  • Elasticized seams and opening

Specifications

Size Large
Material Polyethylene, Polypropylene
Coating Polyethylene
Color Blue
Disposable Single-use
Closure Type Elastic Top and Ankles
Construction Polyethylene, Polypropylene
Seam Type Elastic
Includes Non-woven conductive, dissipative strip
For Use With (Application) Ideal for cleanroom use
Height (English) 7 in.
Height (Metric) 17.78 cm
Product Type Boot Covers
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.