Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Endura™ Premium Cut-Resistant Fitter Gloves

Catalog No. 17401272
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Cut-resistant fitter gloves feature a para-aramid liner for both ANSI Level A5 cut and puncture protection.

  • Blended para-aramid liner for 360° cut protection
  • Heavy-duty premium split-leather is ideal for tough jobs
  • ANSI level 5 puncture resistance
  • Sewn with strong, heat-resistant para-aramid thread for seam integrity
  • Gunn-cut pattern moves seams away from between fingers and toward the back for less rubbing
  • Backs feature leather knuckle straps and elasticized wrists for a snug fit
  • Palms feature 1.4 mm. premium side-split leather

Specifications

Color Gray, Tan
Cut Resistance Rating ANSI A5
Material Leather
Product Type Cut-resistant Gloves
Size Medium
For Use With (Application) Metal fabrication, glass handling, maintenance, automotive, construction, stamping, mining, forestry, steel foundries
Liner Material Aramid Fiber
Model 69BSKFFL/M
Puncture Rating ANSI/ISEA 105-16 Level 5
Series Endura™
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.