Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Superior Glove™ Dexterity™ Cut-Resistant String Knit Gloves, Polyurethane Coating
Click to view available options
Size:
8
9
10
11
12
Description
- Composite fine-gauge knit is highly dexterous
- Cut-resistant, abrasion resistant, puncture resistant
- Polyurethane palm coating provides secure grip and tactility
- Ideal for glass handling, non-marring gloves will not leave residue, smudges or fingerprints
- Dipped over an ergonomic hand form for a natural, comfortable fit
Specifications
Specifications
ANSI Abrasion Rating | ANSI 4 |
Certifications/Compliance (Europe) | CE |
Coating Material | Polyurethane |
Color | Cream, Gray |
Cut Resistance Rating | ANSI A4 |
Material | Aramid Fiber, Silica |
Product Type | Cut-resistant Gloves |
Size | 9 |
EN388 Standard | EN 388:4X31B |
For Use With (Application) | automotive, light Metal stamping, fabrication, small parts handling assembly, Glass handling, sheet Metal work |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction