Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Emerald CX™ Composite Knit Gloves, Nitrile Palm Coating

Catalog No. p-4654423
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Cut-resistant gloves offer a blend of aramid fiber and stainless steel with nitrile palm coating for secure grip during sheet metal work, glass handling, assembly and metal fabrication.

  • 13-gauge blend of aramid fiber and stainless steel
  • Nitrile palm coating provides secure grip
  • High abrasion resistance with comfort and flexibility
  • Nitrile thumb crotch reinforcement to reduce wear in critical areas
  • Touchscreen compatible

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Nitrile
Color Black, Gray
Cut Resistance Rating ANSI A4
Material Aramid Fiber, Stainless Steel Fiber, Nylon
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Metal fabrication, assembly, Glass handling, sheet Metal work
Glove Cuff Style Knit
Model SCXNT/S
Puncture Rating ANSI/ISEA 105-16 Level 4
Series Emerald CX™
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.