Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Emerald CX™ Nylon/Stainless Steel Cut-Resistant Gloves, Foam Nitrile Coating

Catalog No. 17401220
Click to view available options
Size:
6
7
8
9
10
11
12
This item is not returnable. View return policy
This item is not returnable. View return policy

Combination of nylon and fine-gauge steel for cut protection when handling sharp steel or glass.

  • Combination of nylon and fine-gauge steel
  • 13-gauge; cut, abrasion and puncture resistant
  • Extra-long knit wrists protect wrists, keep gloves fitting comfortably
  • Lint-free nylon is lightweight and tough for better dexterity
  • Good touch sensitivity
  • Ideal for handling sharp steel or glass

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Nitrile Foam
Color Black
Cut Resistance Rating ANSI A2
Material Nylon, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size 11
For Use With (Application) HVAC, automotive, sharp Steel, Glass, Metal fabrication, light Metal stamping
Glove Cuff Style Knit
Model S13KBFNT/11
Puncture Rating ANSI/ISEA 105-16 Level 3
Series Emerald CX™
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.