Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Sure Knit™ Dyneema™ Cut-Resistant Gloves

Catalog No. 17401270
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Black
Green
Purple
Red
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Blend of Dyneema™ and stainless steel gloves offer ANSI cut level A5 resistance.

  • Blend of Dyneema™ and stainless steel
  • ANSI cut-level A5
  • Fine 13-gauge knit reduces hand fatigue for all-day comfort and touch sensitivity
  • Treated with AlphaSan™, a silver-based antimicrobial agent that prevents and impedes bacterial growth
  • Anti-microbial agent does not impart any taste to food being handled
  • Touchscreen compatible

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Uncoated
Color Gray
Cut Resistance Rating ANSI A4
Material Dyneema, Stainless Steel Fiber, Spandex
Product Type Cut-resistant Gloves
Size 2X-Large
For Use With (Application) meat processing, food and beverage, handling sharp materials, solar panel manufacturing
Glove Cuff Color Black
Glove Cuff Style Knit
Hem Color Black
Model S13GDSTL/XX
Series Sure Knit™
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.