Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Superior Touch™ Dyneema™ Gloves, Polyurethane Coating

Catalog No. 17400173
Click to view available options
Size:
5
6
7
8
9
10
11
12
This item is not returnable. View return policy
This item is not returnable. View return policy

Cut-resistant gloves with Dyneema™ filament yarn feature a polyurethane coated palm for tacky grip in dry conditions.

  • Polyurethane palm coating is excellent for a 'tacky' grip in dry conditions
  • Lint-free continuous filament yarn
  • High dexterity and flexibility
  • Gloves are washable and can be bleached, at cold temperatures

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A2
Material Dyneema
Product Type Cut-resistant Gloves
Size 7
For Use With (Application) Sharp steel, HVAC, glass metal fabrication, circuit boards, automotive
Glove Cuff Style Knit
Model S13SXGPUQ/7
Puncture Rating ANSI/ISEA 105-16 Level 3
Series Superior Touch™
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.