Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva™ Superloop 150 mL M6 Chromatography Fitting

Catalog No. 01257014 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric) Outer:
45 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer
01-257-014 45 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-257-014 Supplier Cytiva™ Supplier No. 18102385
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in ÄKTA™ Pure Chromatography Systems.

  • Glass tube has a graduated scale of 5 mL increments and seals against the inner end pieces
  • A springloaded valve made from titanium is located in the center of the movable seal
  • When Superloop 150 mL is in loading position, the valve is closed and remains closed during sample loading
  • When the sample is injected onto the column the seal moves downwards so bottom position is reached, the pressure increase causes the valve to open and the buffer passes through the valve
TRUSTED_SUSTAINABILITY

Specifications

Type Fitting
Diameter (Metric) Outer 45 mm
Volume (Metric) 150 mL
Overall Length (Metric) 340 mm
Material Borosilicate Glass
Pressure 14.5 psi
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.