Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperNuclease Recombinant Rabbit Monoclonal Antibody (026), Invitrogen™

Catalog No. PIMA543451
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543451 50 μL
PIMA543096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543451 Supplier Invitrogen™ Supplier No. MA543451
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Specificity: SuperNuclease (Recombinant Serratia marcescens Endonuclease)

SuperNuclease is a genetically-engineered endonuclease. It is a nonspecific nuclease with high activity, capable of completely digesting RNA and DNA (single-stranded, double-stranded, linear, circular and supercoiled forms, that no fewer than five phosphate residues) into 5'-monophosphate-terminated oligonucleotides of 3-5 bases in length. SuperNuclease can be removed by various purification methods.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SuperNuclease
Applications ELISA
Classification Recombinant Monoclonal
Clone 026
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias DNA endonuclease; RNA endonuclease
Host Species Rabbit
Immunogen SuperNuclease (Recombinant Serratia marcescens Endonuclease).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.