Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Superoxide dismutase (SOD) Mn Polyclonal Antibody, QED Bioscience™

Rabbit Polyclonal Antibody
Supplier: Leinco Technologies Inc 13003100UL

This item is not returnable.
View return policy
Description
Superoxide dismutase (SOD) Mn Polyclonal antibody specifically detects Superoxide dismutase (SOD) Mn in Bovine, Canine, Chicken, Drosophila, Guinea Pig, Hamster, Human, Monkey, Mouse, Ovine, Porcine, Rabbit, Rat, Xenopus laevis samples. It is validated for Immunohistochemistry, Western BlotSpecifications
Superoxide dismutase (SOD) Mn | |
Polyclonal | |
Unconjugated | |
SOD2 | |
I79_015272; indophenoloxidase B; IPO B; IPOB; IPO-B; manganese SOD; manganese superoxide dismutase; manganese-containing superoxide dismutase; mangano-superoxide dismutase; manganous superoxide dismutase; mitochondrial superoxide dismutase 2; Mn superoxide dismutase; MNSOD; Mn-SOD; MVCD6; similar to manganous superoxide dismutase; Sod2; Sod-2; sod2.L; sodMt; Superoxide dimutase 2, mitochondrial; superoxide dismutase; superoxide dismutase (Mn type); superoxide dismutase [Mn], mitochondrial; superoxide dismutase 2; superoxide dismutase 2 L homeolog; superoxide dismutase 2, mitochondrial; superoxide dismutase 2, mitochondrial L homeolog | |
Human Mn SOD | |
100 μL | |
Primary | |
Bovine, Canine, Chicken, Drosophila, Guinea Pig, Hamster, Human, Monkey, Mouse, Ovine, Pig, Rabbit, Rat, X. laevis | |
Antibody |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7 | |
P04179, P07895, P09671, P28768, P41976, P41982, P49114, P54712, Q00637 | |
Rabbit | |
Protein A | |
RUO | |
100135623, 100154319, 100760320, 103349944, 104973000, 20656, 24787, 281496, 36878, 374042, 399216, 476258, 6648, 780457 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction