Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superoxide dismutase (SOD) Mn Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502277476
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7476 Supplier Leinco Technologies Inc Supplier No. 13004100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Superoxide dismutase (SOD) Mn Polyclonal antibody specifically detects Superoxide dismutase (SOD) Mn in Bovine, Canine, Chicken, Gerbil, Guinea Pig, Hamster, Human, Monkey, Mouse, Ovine, Porcine, Rabbit, Rat, Xenopus laevis samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot

Specifications

Antigen Superoxide dismutase (SOD) Mn
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7
Gene SOD2
Gene Accession No. P04179, P07895, P09671, P28768, P41976, P41982, P49114, P54712
Gene Alias I79_015272; indophenoloxidase B; IPO B; IPOB; IPO-B; manganese SOD; manganese superoxide dismutase; manganese-containing superoxide dismutase; mangano-superoxide dismutase; manganous superoxide dismutase; mitochondrial superoxide dismutase 2; Mn superoxide dismutase; MNSOD; Mn-SOD; MVCD6; similar to manganous superoxide dismutase; Sod2; Sod-2; sod2.L; sodMt; Superoxide dimutase 2, mitochondrial; superoxide dismutase; superoxide dismutase (Mn type); superoxide dismutase [Mn], mitochondrial; superoxide dismutase 2; superoxide dismutase 2 L homeolog; superoxide dismutase 2, mitochondrial; superoxide dismutase 2, mitochondrial L homeolog
Host Species Rabbit
Immunogen Rat Mn SOD
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100135623, 100154319, 100760320, 103349944, 104973000, 110547884, 20656, 24787, 281496, 374042, 399216, 476258, 6648, 780457
Target Species Bovine, Canine, Chicken, Gerbil, Guinea Pig, Hamster, Human, Monkey, Mouse, Ovine, Pig, Rabbit, Rat, X. laevis
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.