Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Suppressor of Ty 4 homolog 1 Antibody (OTI6B10), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274412G
Description
Suppressor of Ty 4 homolog 1 Monoclonal specifically detects Suppressor of Ty 4 homolog 1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Suppressor of Ty 4 homolog 1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SUPT4H1 | |
Full length recombinant protein of human Suppressor of Ty 4 homolog 1 (NP_003159) produced in E.coli. | |
0.1 mL | |
6827.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI6B10 | |
Western Blot | |
DRB sensitivity-inducing factor 14 kDa subunit, DRB sensitivity-inducing factor small subunit, DSIF p14, DSIF small subunit, SPT4, SPT4HhSPT4, suppressor of Ty (S.cerevisiae) 4 homolog 1, suppressor of Ty 4 homolog 1 (S. cerevisiae), SUPT4H, transcription elongation factor SPT4 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction