Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUPT16H Monoclonal Antibody (OTI3B8)
GREENER_CHOICE

Catalog No. PIMA527204
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527204 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527204 Supplier Invitrogen™ Supplier No. MA527204
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SUPT16H Monoclonal Antibody for Western Blot, IHC (P)

Transcription of protein-coding genes can be reconstituted on naked DNA with only the general transcription factors and RNA polymerase II. However, this minimal system cannot transcribe DNA packaged into chromatin, indicating that accessory factors may facilitate access to DNA. One such factor, FACT, interacts specifically with histones H2A/H2B to effect nucleosome disassembly and transcription elongation. FACT is composed of an 80 kDa subunit and a 140 kDa subunit; this gene encodes the 140 kDa subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUPT16H
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3B8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SUPT16H
Gene Accession No. Q9Y5B9
Gene Alias Cdc68; Chromatin-specific transcription elongation factor 140 kDa subunit; chromatin-specific transcription elongation factor large subunit; DNA unwinding factor 140 kDa subunit; DUF140; facilitates chromatin remodeling 140 kDa subunit; facilitates chromatin transcription complex subunit spt16; FACT; FACT 140 kDa subunit; FACT complex subunit SPT16; FACT complex subunit SPT16-like protein; FACT140; Factp140; FLJ10857; FLJ14010; FLJ34357; fq44d08; HGNC:11465; hSPT16; SPT16; SPT16 homolog, facilitates chromatin remodeling subunit; SPT16/CDC68; suppressor of Ty 16; suppressor of Ty 16 homolog; suppressor of Ty 16 homolog (S. cerevisiae); Supt16; supt16h; wu:fq44d08; zgc:194708; zgc:194732
Gene Symbols SUPT16H
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 187-282 of SUPT16H produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11198
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.