Learn More
Invitrogen™ SUPT16H Monoclonal Antibody (OTI8A1)

Description
SUPT16H Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | SUPT16H |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI8A1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SUPT16H |
| Gene Accession No. | Q9Y5B9 |
| Gene Alias | Cdc68; Chromatin-specific transcription elongation factor 140 kDa subunit; chromatin-specific transcription elongation factor large subunit; DNA unwinding factor 140 kDa subunit; DUF140; facilitates chromatin remodeling 140 kDa subunit; facilitates chromatin transcription complex subunit spt16; FACT; FACT 140 kDa subunit; FACT complex subunit SPT16; FACT complex subunit SPT16-like protein; FACT140; Factp140; FLJ10857; FLJ14010; FLJ34357; fq44d08; HGNC:11465; hSPT16; SPT16; SPT16 homolog, facilitates chromatin remodeling subunit; SPT16/CDC68; suppressor of Ty 16; suppressor of Ty 16 homolog; suppressor of Ty 16 homolog (S. cerevisiae); Supt16; supt16h; wu:fq44d08; zgc:194708; zgc:194732 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.