Learn More
SUPT16H Mouse anti-Human, Clone: OTI3B8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809747

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Transcription of protein-coding genes can be reconstituted on naked DNA with only the general transcription factors and RNA polymerase II. However, this minimal system cannot transcribe DNA packaged into chromatin, indicating that accessory factors may facilitate access to DNA. One such factor, FACT , interacts specifically with histones H2A/H2B to effect nucleosome disassembly and transcription elongation. FACT is composed of an 80 kDa subunit and a 140 kDa subunit; this gene encodes the 140 kDa subunit.Specifications
SUPT16H | |
Monoclonal | |
Unconjugated | |
SUPT16H | |
CDC68, FACTP140, SPT16, SPT16/CDC68 | |
Mouse | |
Affinity Chromatography | |
RUO | |
11198 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3B8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y5B9 | |
SUPT16H | |
Human recombit protein fragment corresponding to amino acids 187-282 of human SUPT16H produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.