Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Supt4h1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568703
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568703 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568703 Supplier Invitrogen™ Supplier No. PA568703
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.

Component of the DRB sensitivity-inducing factor complex (DSIF complex), which regulates mRNA processing and transcription elongation by RNA polymerase II. DSIF positively regulates mRNA capping by stimulating the mRNA guanylyltransferase activity of RNGTT/CAP1A. DSIF also acts cooperatively with the negative elongation factor complex (NELF complex) to enhance transcriptional pausing at sites proximal to the promoter. Transcriptional pausing may facilitate the assembly of an elongation competent RNA polymerase II complex. DSIF and NELF promote pausing by inhibition of the transcription elongation factor TFIIS/S-II. TFIIS/S-II binds to RNA polymerase II at transcription pause sites and stimulates the weak intrinsic nuclease activity of the enzyme. Cleavage of blocked transcripts by RNA polymerase II promotes the resumption of transcription from the new 3' terminus and may allow repeated attempts at transcription through natural pause sites. DSIF can also positively regulate transcriptional elongation and is required for the efficient activation of transcriptional elongation by the HIV-1 nuclear transcriptional activator, Tat. DSIF acts to suppress transcriptional pausing in transcripts derived from the HIV-1 LTR and blocks premature release of HIV-1 transcripts at terminator sequences.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Supt4h1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SUPT4H1
Gene Accession No. 0
Gene Alias AL022777; DRB sensitivity-inducing factor 14 kDa subunit; DRB sensitivity-inducing factor small subunit; DRB sensitivity-inducing factor small subunit 1; DSIF p14; DSIF small subunit; DSIF small subunit 1; hSPT4; small hSpt4 subunit; SPT4; SPT4 homolog, DSIF elongation factor subunit; SPT4H; suppressor of Ty 4 homolog 1; suppressor of Ty 4 homolog 1 (S. cerevisiae); suppressor of Ty 4 homolog 2; suppressor of Ty 4A; Supt4a; Supt4h; Supt4h1; Supt4h1a; Supt4h2; transcription elongation factor SPT4; Transcription elongation factor SPT4 1; transcription elongation factor SPT4-A; zgc:92824; zgc:92824 protein
Gene Symbols SUPT4H1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of rat Supt4h1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287608
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.