Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUPT5H Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040592AF488
Description
SUPT5H Polyclonal antibody specifically detects SUPT5H in Human samples. It is validated for Western BlotSpecifications
SUPT5H | |
Polyclonal | |
Western Blot | |
DRB sensitivity-inducing factor 160 kDa subunit, DRB sensitivity-inducing factor large subunit, DSIF large subunit, DSIF p160, FLJ34157, SPT5hSPT5, SPT5HTat-cotransactivator 1 protein, suppressor of Ty (S.cerevisiae) 5 homolog, suppressor of Ty 5 homolog (S. cerevisiae), Tat-CT1, Tat-CT1 protein, transcription elongation factor SPT5 | |
The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human Suppressor of Ty 5 Homolog using the numbering given in entry NP_003160.2 (GeneID 6829). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
6829 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction