Learn More
SUPT5H Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$167.95 - $630.77
Specifications
| Antigen | SUPT5H |
|---|---|
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1684
|
Bethyl Laboratories, Inc
IHC00210T |
10 μL |
Each for $167.95
|
|
|||||
50-157-1683
|
Bethyl Laboratories, Inc
IHC00210 |
100 μL |
Each for $630.77
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan,
SUPT5H and its binding partner regulate transcriptional elongation by RNA polymerase II. SPT4 and SPT5 are involved in both 5, 6-dichloro-1-beta-D-ribofuranosylbenzimidazole (DRB)-mediated transcriptional inhibition and the activation of transcriptional elongation by the human immunodeficiency virus type 1 (HIV-1) Tat protein.Specifications
| SUPT5H | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Suppressor of Ty 5 homolog (S. cerevisiae), SPT5, SPT5H, Tat-CT1 | |
| Supt5, SUPT5H | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| O00267, O55201 | |
| 20924, 6829 | |
| Between 1040 and C-term | |
| Primary | |
| 4° C | |
| SUPT5H |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.