Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUPT7L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566610
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566610 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566610 Supplier Invitrogen™ Supplier No. PA566610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTMLDIPSEPC SLTIHTIQLI QHNRRLRNLI ATAQAQNQQQ TEGVKTEESE PLPSCPGSPP LPDDLLPLDC KNPNAPFQIR HSD Highest antigen sequence identity to the following orthologs - mouse 94%, rat 93%.

SUPT7L is a protein subunit of the human STAGA complex (SPT3)/TAF9/GCN5 acetyltransferase complex), which is a chromatin-modifying multiprotein complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUPT7L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SUPT7L
Gene Accession No. O94864
Gene Alias 2610524B01Rik; 6030455L14Rik; Adenocarcinoma antigen ART1; KIAA0764; RGD1562206; SPT7 like, STAGA complex gamma subunit; SPT7 like, STAGA complex subunit gamma; SPT7L; SPT7-like STAGA complex gamma subunit; SPTF-associated factor 65 gamma; STAF65; STAF65(gamma); STAF65G; STAF65gamma; STAGA complex 65 gamma subunit; STAGA complex 65 subunit gamma; STAGA complex 65 subunit gamma-like; suppressor of Ty 7 (S. cerevisiae)-like; suppressor of Ty 7-like; SUPT7H; SUPT7L; Unknown (protein for MGC:137593)
Gene Symbols SUPT7L
Host Species Rabbit
Immunogen Recombinant Human SUPT7L. Recombinant protein control fragment (Product #RP-105265).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9913
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.