Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SUPT7L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570808
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 92%; Rabbit: 100%; Rat: 93%.
SUPT7L is a protein subunit of the human STAGA complex (SPT3)/TAF9/GCN5 acetyltransferase complex), which is a chromatin-modifying multiprotein complex.
Specifications
| SUPT7L | |
| Polyclonal | |
| Unconjugated | |
| SUPT7L | |
| 2610524B01Rik; 6030455L14Rik; Adenocarcinoma antigen ART1; KIAA0764; RGD1562206; SPT7 like, STAGA complex gamma subunit; SPT7 like, STAGA complex subunit gamma; SPT7L; SPT7-like STAGA complex gamma subunit; SPTF-associated factor 65 gamma; STAF65; STAF65(gamma); STAF65G; STAF65gamma; STAGA complex 65 gamma subunit; STAGA complex 65 subunit gamma; STAGA complex 65 subunit gamma-like; suppressor of Ty 7 (S. cerevisiae)-like; suppressor of Ty 7-like; SUPT7H; SUPT7L; Unknown (protein for MGC:137593) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 72195 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9CZV5 | |
| SUPT7L | |
| synthetic peptide directed towards the middle region of mouse SUPT7L. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction