Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUPT7L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116758
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116758 Supplier Invitrogen™ Supplier No. PA5116758
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Rat lung Immunogen sequence: TPFPDVMEQV FHEVGIGSVL SLQKFWQHRI KDYHSYMLQI SKQLSEEYER IVNPEKATED AKPVKIKEEP VSDITFPVSE ELEADLASGD QSLPMGVLGA QSERFPSNLE VEASPQASSA EVNASPLWNL AHVKMEPQES EEGNVSGHGV LGSDVFEEPM SGMSEAGIPQ SPDDSDSSYG SHSTDSLMGS S.

SUPT7L is a protein subunit of the human STAGA complex (SPT3)/TAF9/GCN5 acetyltransferase complex), which is a chromatin-modifying multiprotein complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUPT7L
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SUPT7L
Gene Accession No. O94864, Q9CZV5
Gene Alias 2610524B01Rik; 6030455L14Rik; Adenocarcinoma antigen ART1; KIAA0764; RGD1562206; SPT7 like, STAGA complex gamma subunit; SPT7 like, STAGA complex subunit gamma; SPT7L; SPT7-like STAGA complex gamma subunit; SPTF-associated factor 65 gamma; STAF65; STAF65(gamma); STAF65G; STAF65gamma; STAGA complex 65 gamma subunit; STAGA complex 65 subunit gamma; STAGA complex 65 subunit gamma-like; suppressor of Ty 7 (S. cerevisiae)-like; suppressor of Ty 7-like; SUPT7H; SUPT7L; Unknown (protein for MGC:137593)
Gene Symbols SUPT7L
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 210-400 of human SUPT7L (NP_001269658).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313905, 72195, 9913
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.