Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUR1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP234077
Description
SUR1 Polyclonal specifically detects SUR1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
SUR1 | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockdown Validated | |
Q09428 | |
ABCC8 | |
This antibody was developed against a recombinant protein corresponding to amino acids: DHLGKENDVFQPKTQFLGVYFVSSQ | |
0.1 mL | |
ABC Transporters, Lipid and Metabolism, Nutrient Sensing in the Brain | |
6833 | |
Human, Mouse | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ABC36, ATP-binding cassette, sub-family C (CFTR/MRP), member 8, HHF1Sulfonylurea receptor 1, HI, PHHIHRINS, sulfonylurea receptor (hyperinsulinemia), SUR1MRP8, SURATP-binding cassette transporter sub-family C member 8, TNDM2ATP-binding cassette sub-family C member 8 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction