Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUR1 Monoclonal Antibody (N289/16), PerCP
GREENER_CHOICE

Catalog No. PIMA545560
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545560 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545560 Supplier Invitrogen™ Supplier No. MA545560
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45560 was sufficient for detection of SUR1 in 20 μg of mouse brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 160kDa. Does not cross-react with SUR2B. This antibody was formerly sold as clone S289-16.

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies. This protein is a member of the MRP subfamily which is involved in multi-drug resistance. This protein functions as a modulator of ATP-sensitive potassium channels and insulin release. Mutations and deficiencies in this protein have been observed in patients with hyperinsulinemic hypoglycemia of infancy, an autosomal recessive disorder of unregulated and high insulin secretion. Mutations have also been associated with non-insulin-dependent diabetes mellitus type II, an autosomal dominant disease of defective insulin secretion. Alternative splicing of this gene has been observed; however, the transcript variants have not been fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUR1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N289/16
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene ABCC8
Gene Accession No. Q09428, Q09429
Gene Alias ABC36; ABCC8; AM60008PU-N; ATP binding cassette subfamily C member 8; ATP-binding cassette sub-family C member 8; ATP-binding cassette transporter sub-family C member 8; ATP-binding cassette, subfamily C (CFTR/MRP), member 8; ATP-binding cassette, sub-family C (CFTR/MRP), member 8; D930031B21Rik; HHF1; HI; HRINS; LOW QUALITY PROTEIN: ATP-binding cassette sub-family C member 8; MRP8; PH SUR1; PHHI; sulfonylurea receptor; sulfonylurea receptor (hyperinsulinemia); sulfonylurea receptor 1; sulfonylurea receptor subunit 1; sulphonylurea receptor 1; SUR; SUR1; SUR1 protein; SUR1delta2; TNDM2
Gene Symbols ABCC8
Host Species Mouse
Immunogen Fusion protein amino acids 1548-1582 (cytoplasmic C-terminus) of rat SUR1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100764394, 20927, 25559, 6833
Target Species Human, Mouse, Rat, Hamster
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.