Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SUR1 Antibody (S289-16), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259320B
Description
SUR1 Monoclonal specifically detects SUR1 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
SUR1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
ABCC8 | |
Fusion protein amino acids 1548-1582 (cytoplasmic C-terminus) of rat SUR1 | |
100 μL | |
Primary | |
Detects 160kDa. Does not cross-react with SUR2B. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S289-16 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ABC36, ATP-binding cassette, sub-family C (CFTR/MRP), member 8, HHF1Sulfonylurea receptor 1, HI, PHHIHRINS, sulfonylurea receptor (hyperinsulinemia), SUR1MRP8, SURATP-binding cassette transporter sub-family C member 8, TNDM2ATP-binding cassette sub-family C member 8 | |
Mouse | |
Protein G purified | |
ABC Transporters, Lipid and Metabolism, Nutrient Sensing in the Brain | |
6833 | |
Human, Mouse, Rat, Hamster | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction