Learn More
Description
This product may be supplied with a high degree of hydration and some residual NaCl, the amounts of which are batch dependent. Please refer to the Certificate of Analysis to obtain the batch specific Net Product Content.
Specifications
Specifications
| Chemical Name or Material | 8,8'-[Carbonylbis[imino-3,1-phenylenecarbonylimino(4-methyl-3,1-phenylene)carbonylimino]]bis-1,3,5-naphthalenetrisulfonic acid hexasodium salt |
| CAS | 129-46-4 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Target | P2X Receptor Antagonists |
| Molecular Formula | C51H34N6Na6O23S6 |
| Purity | 0.98 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
