Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chembio SURE CHECK™ HIV 1/2 Assay
GSA_VASDP

Catalog No. 23999105
Change view
Click to view available options
Quantity:
25 test devices per kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-999-105 25 test devices per kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-999-105 Supplier Chembio Supplier No. 6095070
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • 25 Foil Pouches with Test Device and Buffer Vial attached, Bandage, Safety Lancet and Desiccant inside
  • 25 Disposable Test Stands
  • 25 Subject Information Sheets
  • 1 Quick Reference Guide
  • 1 Product Insert

Unique, easy-to-use, self-contained collection and testing device for the rapid, visual detection of antibodies to HIV-1 and HIV-2 in whole blood, serum or plasma.

  • Rapid results in 15 minutes
  • Self-contained collection and testing
  • Detects HIV 1/2 antibodies
  • Serum, plasma, whole blood samples
  • Each test device includes running buffer, bandage, sterile safety lancet and disposable stand.

Certifications

FDA PMA BP050009 / 0; CLIA Waived for Fingerstick and Venous Whole Blood

Warranty and Services

Until expiration date printed on test pouch and kit box

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 8°C to 30°C
For Use With (Application) Detection of antibodies to HIV 1/2 in serum, plasma or whole blood
Includes 25 Foil Pouches with Test Device and Buffer Vial attached, Bandage, Safety Lancet and Desiccant inside
25 Disposable Test Stands
25 Subject Information Sheets
1 Quick Reference Guide
1 Product Insert
Quantity 25 test devices per kit
Sample Size 2.5 μL requirement
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.