Learn More
Fisher Healthcare™ Sure-Vue™ Fecal Occult Blood Slide Test System
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 Pack
1000 Pack
20 Pack
80 Pack
Type:
FOBT Developer
FOBT Developer (extra sen)
FOBT Single Slide (extra sen)
FOBT Single Slide Test
FOBT Take Home Kit
FOBT Take Home Kit (extra sen)
Description
Available in traditional guaiac format for routine screening applications and extra-sensitive format for more stringent testing
- Take-Home Kits include one FOBT triple slide with on-slide monitors, one mailing envelope, three applicator sticks, tissues and instructions
- Use single slides when bleeding in the gastrointestinal tract is strongly suspected as in persons with symptoms of ulcers, black stools or in post-operative patients
- Use triple slides for serial testing as recommended by the American Cancer Society for screening of asymptomatic patients
- Bottles of developer for use with Fecal Occult Blood Slide Test Systems
- 23-038-033 is for use with normal slides
- 23-038-037 is for use with extra-sensitive slides
For screening purposes only
Specifications
Specifications
| Clia Complexity | Waived |
| Type | FOBT Take Home Kit (extra sen) |
| For Use With (Application) | Detection of fecal occult Blood |
| Includes | 80 Extra-sensitive take home kits (each contains triple slide with monitors, collection tissues, applicator, test instructions and mailing envelope) and 6 x 15mL bottles of extra-sensitive developer. |
| Quantity | 80 Pack |