Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Healthcare™ Sure-Vue™ Signature Control Set, Cryptosporidium/Giardia
Shop All Fisherbrand Products
Click to view available options
Detectable Analytes:
Cryptosporidium Oocyst Antigen, Giardia Cyst Antigen
Description
- One test that detects and differentiates between Cryptosporidium and Giardia
- Cryptosporidium CPT Code - 87328, Giardia CPT Code - 87329
- Proven Performance with high sensitivity (Crypto - 97%/GI - 100%) and specificity (Crypto - 99.5%/GI - 99.1%)
- Will improve turnaround time for test results in approximately 10 minutes
Specifications
Specifications
| Content And Storage | 2°C to 8°C (35.6°F to 46.4°F) |
| CPT Code | Cryptosporidium 87328, Giardia 87329 |
| Clia Complexity | Not Applicable |
| Detectable Analytes | Cryptosporidium Oocyst Antigen, Giardia Cyst Antigen |
| Format | Vial |
| Sample Type | Stool |
| Sensitivity | Crypto 97%, GI 100% |
| Type | Control Kit |
| Includes | 1 mL Crypto, 1 mL Giardia |
| Packaging Type | Test Kit |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction