Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ Signature Influenza A and B Test Kit

Catalog No. 23-200-279 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 Tests
This item is not returnable. View return policy
This item is not returnable. View return policy

Sure-Vue™ Signature Influenza A and B Test Kit for rapid detection of influenza type A and B nucleoprotein antigens directly from nasal and nasopharyngeal swab specimens from patients. High sensitivity and specificity allow for accurate diagnostic testing that is efficient and cost-effective.

  • Results in 10 minutes
  • High performance - near or exceeding that of traditionally more sensitive reader devices without the need for an instrument
  • Sensitivity for flu A: 90.3%; flu B: 88%
  • Specificity for flu A: 96.7%; flu B: 99.2%
  • Performance characteristics for influenza A were established during the U.S. 2018-2019 influenza season when A/H1N1pdm09 and influenza A/H3N2 were the predominant influenza A viruses in circulation, and the influenza B Yamagata and Victoria lineages were in co-circulation
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CLIA waived, 510k approved
Content And Storage Room Temperature (15°C to 30°C, 59°F to 86°F)
CPT Code 87804QW (Flu A), 87804QW-59 (Flu B)
Clia Complexity Waived (Nasal and NP Swab)
Detectable Analytes Influenza A and B
Detection Method Influenza antigens from nasal or NP swabs
DoA Calibrators No
Format Manual Dipstick
Sensitivity A: 90.3% B: 88.0%
Type Rapid Test
For Use With (Application) Lateral Flow Rapid Test
Includes 25 test sticks, 25 nasal swabs, 25 extraction buffer vials, Influenza A+ Control Swab, Influenza B+ Control Swab, IFU, QRG, Workstation
Packaging Type Test Kit
Product Line Sure-Vue™ Signature
Quantity 25 Tests
Specificity A: 96.7%, B: 99.2%
Test Time 10 min.
Analysis Time 10 min.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.