Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ SureOne™ Aerosol Barrier Pipette Tips
GREENER_CHOICE

Catalog No. p-8308982 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
2 to 20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
02-707-435 2 to 20 μL
02-707-456 0.1 to 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 02-707-435
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Choose from a comprehensive line of pipette tips for demanding liquid-handling applications.

Choose from a comprehensive line of pipette tips for demanding liquid-handling applications

Universal fit for research grade pipettes.
  • Engineered for increased accuracy and are compatible with single-channel, multichannel, and electronic pipettes
  • DNase-/RNase-free; sterile tips are also pyrogen-free
  • Sterile and clear color
  • Filter eliminates aerosol cross-contamination without sealing or trapping samples
  • Tips allow for noncontaminated sample recovery
Not autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 2 to 20 μL
Filtered Filtered
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Length (English) 2 in.
Length (Metric) 5.08cm
Format Hinged Rack
Tip Style Micro Point
For Use With (Equipment) Eppendorf
Product Line SureOne
Show More Show Less

As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.