Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SurePac™ Bio 550 SEC MDi™ HPLC Columns

Catalog No. 43903152131
Change view
Click to view available options
Length (Metric):
30 mm
150 mm
Diameter (Metric):
2.1 mm
4.6 mm
7.8 mm
Column Format:
Analytical Column
Guard Column
Flow Rate:
0.6 mL/min.
1.0 mL/min.
125 μL/min.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Column Format Flow Rate
43-903-152131 150 mm 2.1 mm Analytical Column 125 μL/min.
43-903-154631 150 mm 4.6 mm Analytical Column 0.6 mL/min.
43-903-157831 150 mm 7.8 mm Analytical Column 1.0 mL/min.
43-903-032131 30 mm 2.1 mm Guard Column 125 μL/min.
43-903-034631 30 mm 4.6 mm Guard Column 0.6 mL/min.
43-903-037831 30 mm 7.8 mm Guard Column 1.0 mL/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 43-903-152131 Supplier Thermo Scientific™ Supplier No. 43903152131
Only null left
Add to Cart
Add to Cart

Experience the future of Size-Exclusion Chromatography (SEC) with SurePac™ Bio 550 SEC MDi™ analytical columns and guards exclusively designed to deliver robust and reproducible performance at high resolution and throughput to accelerate your therapeutics analysis process.

Experience the future of Size-Exclusion Chromatography (SEC) with SurePac™ Bio 550 SEC MDi™ analytical columns and guards exclusively designed to deliver robust and reproducible performance at high resolution and throughput to accelerate your therapeutics analysis process. The column's groundbreaking MDi™ technology integrates our advanced 3 μm monodisperse silica particle platform with bioinert hardware to ensure superior reliability for best sample recovery. SurePac™ Bio 550 SEC MDi™ column is ideal for AAV gene therapeutics analysis and offers excellent selectivity due to the dedicated smaller particle (3 μm) size combined with wide-pore (550Å) for separating monomeric AAVs from aggregates based on their size differences.

SurePac Bio 550 SEC MDi columns and guards are based on high-purity, spherical, porous (550 Å), 3 μm monodispersed silica particles that are covalently modified with a proprietary diol hydrophilic layer. This unique design yields shorter diffusion distances and enhanced reproducible mass transfer and narrower peaks. The consistent size of these particles facilitates precise control over media synthesis and column packing, significantly improving column-to-column and lot-to-lot reproducibility. The columns are housed in a hydrophilic bioinert-coated stainless-steel hardware to eliminate secondary interactions, ensuring optimal performance during the initial injection process.

  • Ideal for AAV gene therapy characterization and quality control assessment due to selectivity of the dedicated smaller particle (3 μm) combined with wide-pore (550Å) size
  • Superior column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing
  • High-efficiency peaks with excellent recovery
  • Higher resolution and throughput for fast turnaround time without sacrificing performance
  • Bioinert hardware to eliminate analyte absorption
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 2.1 mm
Flow Rate 125 μL/min.
Length (Metric) 150 mm
Max. Pressure 3500 psi (241 bar)
Particle Size 3 μm
pH 2 to 8
Quantity 1 Ea.
Temperature 20°C to 60°C
Stationary Phase Proprietary
Packing Material Monodisperse Silica Resin
Phase Size Exclusion
Technique SEC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.