Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SurePac™ Oligo RP MDi™ HPLC Columns

Catalog No. 43712022132
Change view
Click to view available options
Diameter (Metric):
2.1 mm
300 μm
Length (Metric):
20 mm
50 mm
100 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric)
43-712-022132 2.1 mm 20 mm
43-712-052132 2.1 mm 50 mm
43-712-102132 2.1 mm 100 mm
43-712-050332 300 μm 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 43-712-022132 Supplier Thermo Scientific™ Supplier No. 43712022132
Add to Cart
Add to Cart

SurePac Oligo RP MDI is a robust reversed-phase column designed for oligonucleotide analysis by HPLC and LC-MS. It delivers high recovery, bioinert column hardware, and reproducible separations, enabling reliable profiling of oligonucleotides of any size.

Thermo Scientific™ SurePac™ Oligo RP MDi™ HPLC columns deliver high-resolution, reproducible separations for therapeutic oligonucleotides and nucleic acids across R and D and QC workflows. Built on monodisperse, supermacroporous polymeric packing with bio-inert hardware, they provide stable performance even under elevated pH and temperature conditions, resolving truncation impurities while supporting large oligos and intact nucleic acids for scalable LC-MS and LC-UV analysis.

  • Purpose-built for oligonucleotide RP separations
  • Optimized for DNA and RNA oligonucleotides, including modified sequences, delivering consistent retention and selectivity across short and long oligos.
  • Bioinert, metal-free fluid path
  • Reduces metal/oligonucleotide interactions to improve recovery, minimize adsorption, and deliver cleaner peak shapes for sensitive analyses.
  • Optimized for ion-pair RP LC-MS
  • Fully compatible with common ion-pairing reagents such as HFIP/TEA, enabling sensitive, reproducible LC-MS detection.
  • High recovery of long and modified oligonucleotides
  • High resolution power for oligonucleotides spanning over a wide size range from small to (ultra) large
  • Supports efficient analysis of long, hydrophobic, and chemically complex oligos without compromising signal or resolution.
  • Robust stationary phase for demanding methods
  • Maintains stable performance under high organic content and elevated temperatures, supporting fast gradients and long column lifetimes.
  • Excellent peak shape and impurity resolution
  • Delivers sharp, symmetrical peaks and reliable separation of closely related species, including impurities and sequence variants.
  • Reproducible, manufacturing-controlled performance
  • Ensures consistent results across columns and batches, simplifying method transfer and routine use.
  • Flexible for development and routine workflows
  • Suitable for method development, impurity profiling, and QC, reducing the need for multiple column chemistries.
  • Compatible with UHPLC and HPLC systems
  • Designed for seamless integration into modern LC platforms and existing laboratory setups.
  • Built for confident bioanalytical results
  • Improves robustness, reduces variability, and supports reliable data generation in oligonucleotide workflows.
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical
Diameter (Metric) 2.1 mm
Flow Rate 0.2 to 1 mL/min.
Length (Metric) 20 mm
Max. Pressure 6500 psi (448 bar)
Particle Size 2.5 μm
pH 2 to 12
Pore Size Proprietary supermacroporous
Temperature 5°C to 90°C
Stationary Phase Poly-DVB
Packing Material Polymeric
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.