Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SureQuant™ AKT Pathway (Phospho) Multiplex Panel (Absolute Quantitation)

Catalog No. pia40084
Click to view available options
Quantity:
10 reactions kit

SureQuant AKT pathway multiplex assays are validated, modular reagents for protein quantitation using mass spectrometry (MS). Together, the IP and MS Sample Preparation Module and Absolute or Relative Quantitation Modules provide a complete solution for analyzing proteins in the AKT/mTOR pathway.

The SureQuant Quantitation Modules contain 30 unique peptides from 10 AKT-mTOR signaling pathway target proteins for multiplexed quantitation of unique peptides using LC and MS. SureQuant kits utilize multiplex immunoprecipitation coupled with mass spectrometry for simultaneous enrichment and quantitation of target proteins in the AKT/mTOR signaling pathway. Each multiplex panel contains two modules: 1) the IP and MS Sample Preparation Module, which includes all the reagents necessary to immunoenrich target proteins with subsequent in-solution MS sample preparation, and 2) an Absolute or Relative Quantitation Module, including a system suitability standard, and HeavyPeptide™ and LightPeptide™ AQUA Ultimate mixtures.

For relative quantitation, a HeavyPeptide AQUA Ultimate mixture is provided for use as a spike-in internal standard in the multiplexed sample. For absolute quantitation, HeavyPeptide and LightPeptide AQUA Ultimate mixtures are provided with the Pierce™ 6 Protein Digest are used to generate a standard curve in a matrix. The Pierce™ LC-MS/MS System Suitability Standard (7 x 5 mix) is included in both modules to assess the dynamic range of LC-MS/MS systems before running the calibration curves and samples.

The IP and MS Sample Preparation Module enables highly effective antigen IP and co-IP for MS analysis. Cells are first lysed and incubated with an IP-MS–verified biotinylated antibody mixture. The antibody–antigen complex is then captured on MS-compatible streptavidin magnetic beads, and nonspecific proteins are removed through washing with MS-compatible buffers. The target proteins are eluted using trypsin and are compatible with MS sample preparation. The MS sample preparation has been streamlined to be completed in 4 hours so that the sample can be analyzed by MS on the same day.

The AKT/mTOR signaling pathway serves as a master regulatory pathway for cell growth and proliferation and plays a vital role in the signaling cascade. Aberrant activation of this pathway promotes survival of tumor cells in many types of cancer. SureQuant kits are ideal for monitoring AKT/mTOR expression levels, isoforms, and phosphorylation status in a single assay. The ability to simultaneously monitor and quantitate in a single sample is a powerful tool for assessment of normal and abnormal cell growth.

SureQuant AKT Pathway Multiplex Panels are:
Complete—includes all reagents for successful monitoring of MS system performance, sample preparation, and quantitation of target proteins and peptides
Verified—antibodies, peptides, and control lysate are rigorously tested for specificity and successful quantitation of each target peptide
Multiplex—able to quantitate different targets from the AKT/mTOR pathway, RAS, and p53 isoforms
Flexible—modular format allows for immune-enrichment only, or in combination with relative or absolute quantitation

TRUSTED_SUSTAINABILITY

Specifications

Description SureQuant™ AKT Pathway (Phospho) Multiplex Panel (Absolute Quantitation)
Detection Method Mass Spectrometry
For Use With (Equipment) Thermo Scientific™ Q Exactive Mass Spectrometer, Mass Spectrometer
Shipping Condition Approved for shipment on Wet or Dry Ice
Workflow Step Target Selection
Product Line SureQuant™
Quantity 10 reactions kit
For Use With (Application) Mass Spectrometry
Type Multiplex Panel
Final Product Type Peptide
Starting Material Protein samples
Sufficient For 10 Reactions
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.