Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SureQuant™ IP-MS Sample Preparation Kits

Catalog No. p-7225600
Click to view available options
Description:
Protein A/G Magnetic Beads
Streptavidin Magnetic Beads

Thermo Scientific SureQuant IP-MS Sample Preparation Kits provide an optimized solution for the immunoprecipitation and MS peptide preparation of samples upstream of LC-MS/MS analysis.

Thermo Scientific SureQuant IP-MS Sample Preparation kits (Protein A/G or streptavidin) provide an optimized solution for the immunoprecipitation and MS peptide preparation of samples upstream of LC-MS/MS analysis. These kits have been rigorously verified using numerous multiplexed target antigens with varying expression levels. The SureQuant product family enables the use of the immunoprecipitation (IP) sample prep kits and peptide quantitation modules together or independently for SRM/PRM analysis.

Complete—kits include reagents for successful immunoprecipitation and MS peptide preparation for target proteins for SRM/PRM analysis
Multiplex—IP-MS kits have been designed for multiplex IP using both Protein A/G and Streptavidin magnetic beads
Flexible—universal reagents enable individualized selection of targets for IP to be used alone or combined with SureQuant peptide quantitation panels
Optimized—simplified and condensed procedure to prepare samples for LC-MS analysis
Low background—binding, wash, and elution buffers optimized to remove contaminants and minimize enrichment of background proteins

The SureQuant Protein A/G and Streptavidin IP-MS Sample Preparation kits have been designed to support targeted quantitation using SureQuant or SRM/PRM analysis. The kits contain high-quality Pierce Protein A/G or Streptavidin magnetic beads together with reagents that have been optimized for MS compatibility. The streamlined procedure enables digestion immediately after the immunoprecipitation elution step, facilitating MS sample preparation in ∼4 hours for same-day LC-MS/MS analysis.

Applications:
• Enrichment of low abundance targets to improve MS sensitivity
• Identification of target proteins that are not detectable by western blotting
• Quantitation of target proteins by LC-MS analysis
• Screening of antibodies for IP-MS
• Enrichment of target proteins upstream of targeted analysis (SRM)

TRUSTED_SUSTAINABILITY

Specifications

Description Protein A/G Magnetic Beads
Kit Contents Trypsin Protease, 2 x 20 μg
Trypsin Storage Solution, 250 μL
Protein A/G Magnetic Beads, 1 mL
IP-MS Cell Lysis Buffer, 100 mL
IP-MS Wash Buffer A, 75 mL
IP-MS Wash Buffer B, 40 mL
IP Elution and MS Sample Prep Buffer, 10 mL
Alkylation Solution, 1 mL/vial
Low Protein-Binding Collection Tubes, 50 x 1.5 mL
Bond-Breaker™ TCEP Solution, Neutral pH, 0.5 mL
10% Pierce™ Trifluoroacetic Acid (TFA), Sequencing Grade, 0.5 mL
1M MgCl2, 0.75 mL
For Use With (Equipment) Mass spectrometer
Product Line SureQuant™
Content And Storage Store trypsin at 4°C.
Store trypsin storage solution at -20°C.
Starting Material Cells, Tissue
Detection Method Mass Spectrometry
Workflow Step Sample prep enrichment
Quantity 20 Reactions
For Use With (Application) MS-compatible immunoprecipitation
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.