Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SureQuant™ Phosphopeptide Suitability Standards

Catalog No. p-7228833
Click to view available options
Concentration:
100 fmol/μL
500 fmol/μL

SureQuant Phosphopeptide Standards enable assessment of LC-MS/MS system suitability for phosphopeptide analysis and multiplexed quantitation of phosphorylation of 89 key target proteins from 7 signaling pathways using LC-MS/MS analysis.

The SureQuant Multipathway Phosphopeptide Standard contains an optimized mixture of 131 isotopically labeled phosphopeptides to simultaneously monitor and quantitate phosphorylation signaling. The SureQuant Phosphopeptide Suitability Standard contains a mixture of 20 isotopically labeled phosphopeptides with increasing hydrophobic properties for LC optimization and monitoring of LC-MS/MS system performance. When used together, the standards help ensure optimal instrument performance and sample assessment for phosphorylation events.

The specific phosphopeptides in the SureQuant Multipathway Phosphopeptide Standard have been chosen to cover biologically relevant phosphorylation sites for 89 key proteins from seven different signaling pathways including EGFR/HER, RAS-MAPK, PI3K/AKT/mTOR, AMPK, death/apoptosis, and stress (p38/SAPK/JNK). For optimal results, MS sample preparation should include phosphopeptide enrichment, with the standard spiked in before MS sample preparation (1 pmol) or after phosphopeptide enrichment (0.8 pmol). The ability to simultaneously monitor and quantitate these targets in a single sample is a powerful tool for assessment of normal and abnormal signaling pathway activity.

The phosphopeptides selected for the SureQuant Phosphopeptide Suitability Standard represent relevant proteins from key signaling pathways that include serine, threonine, and tyrosine phosphorylation. The 20 phosphopeptides have a wide range of retention times and have been designed for use with phosphopeptide samples to assess system performance and to better enable prediction of peptide retention times to maximize phosphopeptide identifications.

Features include:
• Optimized—validated mixture of 131 HeavyPeptide AQUA Basic phosphorylated peptides (SureQuant Multipathway standard) or 20 HeavyPeptide AQUA Basic phosphopeptides (SureQuant Suitability standard)
Multiplex—suitable for relative quantitation of phosphorylationevents for 89 key proteins in seven cellular signaling pathways (SureQuant Multipathway standard)
Predictive—range of peptides with increasing hydrophobic properties for method optimization and calculation of peptide retention times
Monitoring—excellent QC assay tool for quality assessment of LC and MS instrument performance (SureQuant Suitability standard)
Convenient—provided in a ready-to-use liquid format

Applications:
• Standard for data-dependent acquisition (DDA)
• Standard for targeted MS (e.g., SRM, PRM)
• Optimization of LC parameters
• Identification of total peptide elution window
• Optimization of MS parameters

For optimal results, MS sample preparation should include phosphopeptide enrichment. We recommend use of the EasyPep Maxi MS Sample Prep Kit (Cat. No. A45734) combined with the High Select Phosphopeptide Enrichment Kits (Cat. No. A32992, A32993, or A52283) for sample preparation. Standardized methods and data process templates are provided for streamlined data acquisition and analysis.

TRUSTED_SUSTAINABILITY

Specifications

Description SureQuant™ Multipathway Phosphopeptide Standard (100 fmol/μL)
Concentration 100 fmol/μL
Quantity 200 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.