Learn More
Thermo Scientific™ SureQuant™ Protein A/G IP-MS Sample Preparation Kit, 20 Reactions PROMO
Description
- Complete—kits include reagents for successful immunoprecipitation and MS peptide preparation for target proteins for SRM/PRM analysis
- Multiplex—IP-MS kits have been designed for multiplex IP using both Protein A/G and Streptavidin magnetic beads
- Flexible—universal reagents enable individualized selection of targets for IP to be used alone or combined with SureQuant peptide quantitation panels
- Optimized—simplified and condensed procedure to prepare samples for LC-MS analysis
- Low background—binding, wash, and elution buffers optimized to remove contaminants and minimize enrichment of background proteins
The SureQuant Protein A/G and Streptavidin IP-MS Sample Preparation kits have been designed to support targeted quantitation using SureQuant or SRM/PRM analysis. The kits contain high-quality Pierce Protein A/G or Streptavidin magnetic beads together with reagents that have been optimized for MS compatibility. The streamlined procedure enables digestion immediately after the immunoprecipitation elution step, facilitating MS sample preparation in 4 hours for same-day LC-MS/MS analysis.
Applications:
- Enrichment of low abundance targets to improve MS sensitivity
- Identification of target proteins that are not detectable by western blotting
- Quantitation of target proteins by LC-MS analysis
- Screening of antibodies for IP-MS
- Enrichment of target proteins upstream of targeted analysis (SRM)
Specifications
Specifications
Kit Contents | Trypsin Protease, 2 x 20 μg, Trypsin Storage Solution, 250 μL, Protein A/G Magnetic Beads, 1 mL, IP-MS Cell Lysis Buffer, 100 mL, IP-MS Wash Buffer A, 75 mL, IP-MS Wash Buffer B, 40 mL, IP Elution and MS Sample Prep Buffer, 10 mL, Alkylation Solution, 1 mL/vial, Low Protein-Binding Collection Tubes, 50 x 1.5 mL, Bond-Breaker™ TCEP Solution, Neutral pH, 0.5 mL, 10% Pierce™ Trifluoroacetic Acid (TFA), Sequencing Grade, 0.5 mL, 1M MgCl2, 0.75 mL |
For Use With (Equipment) | Mass Spectrometer |
Product Line | SureQuant™ |
Content And Storage | Store trypsin at 4°C. Store trypsin storage solution at -20°C. |
Starting Material | Cells, Tissue |
Detection Method | Mass Spectrometry |
Workflow Step | Sample prep enrichment |
Quantity | 20 Reactions |
For Use With (Application) | MS-compatible immunoprecipitation |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.