Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SureQuant™ RAS Isoform Relative Quantitation Module

Catalog No. pia40097
Click to view available options
Quantity:
10 reactions kit

Thermo Scientific™ SureQuant™ RAS Isoform Relative Quantitation Module provides a complete quantitation solution for the analysis of key RAS expression using mass spectrometry (MS).

  • Complete - contains validated AQUA HeavyPeptide mix, diluent, matrix, and system suitability standard including optimized acquisition and analysis methods
  • Validated - peptides rigorously tested for successful quantitation of each target peptide
  • Multiplex - able to quantitate NRAS, KRAS, and HRAS isoforms
TRUSTED_SUSTAINABILITY

Specifications

Detection Method Mass Spectrometry
For Use With (Equipment) Thermo Scientific™ Q Exactive Mass Spectrometer, Mass Spectrometer
Shipping Condition Approved for shipment on Wet or Dry Ice
Workflow Step Target Selection
Product Line SureQuant™
Quantity 10 reactions kit
For Use With (Application) Mass Spectrometry
Type Quantitation Module
Final Product Type Peptides
Starting Material Protein Samples
Sufficient For 10 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.