Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SureTect™ Cronobacter species PCR assay

Catalog No. A56845 Shop All Thermo Scientific Products
This item is not returnable. View return policy
Catalog No. A56845 Supplier Thermo Scientific™ Supplier No. A56845
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Simply and accurately detect Cronobacter species in low or large sample sizes of powdered infant formula, and related ingredients, and production environment samples in less than 20 hours with the Thermo Scientific™ SureTect™ Cronobacter Species PCR Assay.

Simply and accurately detect Cronobacter species in low or large sample sizes of powdered infant formula, and related ingredients, and production environment samples, in less than 20 hours with the Thermo Scientific SureTect Cronobacter Species PCR Assay.

The assay is AFNOR (ISO 16140-2:2016) AOAC (RI) validated on infant formula and cereals with and without probiotics, related ingredients, and production environment samples.

The new SureTect assay format incorporates color-coded plastics with orientation markings and a breakable strip format, colored reagents and pierceable seals to enable an optimized workflow for maximum laboratory efficiency and productivity.

The SureTect System is an optimized solution combining speed and performance in an easy-to-use, cost-effective platform—giving you results you can trust. SureTect Cronobacter Species PCR Assay protocols are designed to be simple while producing repeatable, accurate results for production environment samples and powdered infant formula (with or without probiotics) and ingredients.

Simple

  • Easy-to-use with single enrichment step and pre-dispensed reagents
  • Target-specific colored assay plastics
  • Clearly oriented lysis and assay trays
  • Color indicator in lysis reagent reducing possibility for human error

Fast and Flexible

  • Option to automate the lysis and PCR sample preparation
  • Optimized workflow delivers on-screen results in as little as 20 hours
  • Maximum flexibility for operational efficiency
  • Run all assays simultaneously in the same run
  • Test up to 96 samples in each run
  • Choice of pierceable seals or caps
  • Detect multiple targets in the same run with universal PCR protocols across the range

Smart

  • Intuitive, user-friendly software delivers reliable results, with no subjective interpretation
  • Actionable data supports fast decisions in a complex product supply chain
  • Internal positive control (IPC) in every assay for complete confidence in results
  • NOW available with SureClean™ for your most challenging samples where free DNA can influence result performance

The Thermo Scientific SureTect Cronobacter Species PCR Assay Kit includes:

  • Lysis Reagent 1 Tubes
  • Proteinase K
  • Cronobacter PCR Assay Tubes
  • ThermoScientific 96-Well Pierceable Seals
  • PCR Caps

The Thermo Scientific Suretect pathogen detection assays are validated to run on Applied Biosystems™ QuantStudio™ 5 Food Safety Real Time PCR instrument with RapidFinder™ Analysis Software v3.0 (or later) and the Applied Biosystems 7500 Fast Food Safety 96-well Real-Time PCR instrument using the RapidFinder™ Express Software v2.0 (or later).

TRUSTED_SUSTAINABILITY

Specifications

Product Line SureTect
Target Organism Class Cronobacter species
For Use With (Application) Food and Beverage
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.