Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SureTect™ Escherichia coli O157:H7 PCR Assay

Catalog No. A56844 Shop All Thermo Scientific Products
Change view
Click to view available options
Target Organism Class:
E. coli O157:H7
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Target Organism Class
A56844 E. coli O157:H7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A56844 Supplier Thermo Scientific™ Supplier No. A56844
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Simply and accurately detect E. coli O157:H7 on beef meats, dairy, fruits, and vegetables including large test portion sizes with the Thermo Scientific™ SureTect™ E. coli O157:H7 PCR Assay.

Detect E. coli O157:H7 rapidly with the highly specific, real-time Thermo Scientific™ SureTect™ E. coli O157:H7 PCR Assay.

The assay holds AOAC Performance Tested MethodsSM approval granted by the AOAC Research Institute for analysis of up to 375 g of raw beef meats and fresh produce. The assay is also validated according to ISO 16140-2:2016 and certified by NF Validation™ by AFNOR for analysis of dairy products and up to 375 g raw meat products and vegetables.

The new SureTect assay format incorporates color-coded plastics with orientation markings and breakable strip format, colored reagents, and pierceable seals to enable an optimized workflow for maximum laboratory efficiency and productivity.

The SureTect System is an optimized solution combining speed and performance in an easy-to-use, cost-effective platform—giving you results you can trust.

Simple

  • Easy-to-use with single enrichment step and pre-dispensed reagents
  • Target-specific, colored assay plastics
  • Clearly oriented lysis and assay trays
  • Color indicator in lysis reagent reducing possibility for human error

 Fast and Flexible

  • Option to automate the lysis and PCR sample preparation
  • Optimized workflow delivers on-screen results in as little as 10-22 hours
  • Maximum flexibility for operational efficiency
  • Run all assays simultaneously in the same run
  • Test up to 96 samples in each run
  • Choice of pierceable seals or caps
  • Detect multiple targets in the same run with universal PCR protocols across the range

 Smart

  • Intuitive, user-friendly software delivers reliable results, with no subjective interpretation
  • Actionable data supports fast decisions in a complex product supply chain
  • IPC in every assay for complete confidence in results

The Thermo Scientific SureTect E. coli O157:H7 PCR Assay Kit includes:

  • Lysis Reagent 1 Tubes
  • Proteinase K
  • E. coli O157:H7 PCR Tubes
  • ThermoScientific 96-Well Pierceable Seals
  • PCR Caps

Enriched samples are pipetted into pre-filled lysis tubes, along with Proteinase K, before incubation, to lyse any bacterial cells present in the sample and release their DNA into the solution. The lysate is then loaded into the SureTect E. coli O157:H7 PCR tube to re-hydrate the pre-loaded PCR pellet which contains all of the necessary components and reagents for PCR, including a probe, primers, and DNA template for the internal positive control (IPC). 

The PCR tubes are then sealed, loaded into the Applied Biosystems™ QuantStudio™ 5 Food Safety Real Time PCR instrument using RapidFinder™ Analysis Software v3.0 (or later) or the Applied Biosystems 7500 Fast Food Safety 96-well Real-Time PCR instrument using the RapidFinder™ Express Software v2.0 or later. After completing the run, interpreted results are clearly displayed and can be reported, stored, printed, or downloaded as required. The internal amplification control confirms correct assay conditions have occurred.

TRUSTED_SUSTAINABILITY

Specifications

Product Line SureTect
Target Organism Class E. coli O157:H7
For Use With (Application) Food and Beverage
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.