Learn More
Thermo Scientific™ SureTect™ Salmonella Infantis PCR Assay
Description
Rapidly detect Salmonella Infantis in carcass rinses and poultry part samples with the Thermo Scientific™ SureTect™ Salmonella Infantis PCR Assay workflow.
Rapidly detect Salmonella Infantis in carcass rinses and in up to 375 g poultry parts samples with the Thermo Scientific™ SureTect™ Salmonella Infantis PCR Assay, which incorporates optimized primers and probe components for highly specific results.
Designed specifically for Salmonella Infantis control programs in poultry production, the SureTect Salmonella Infantis PCR Assay has been extensively tested and verified on poultry matrices from food manufacturing environments and addresses process challenges by offering accuracy and efficiency in product release or intervention.
Combine Thermo Scientific™ SureTect™ Salmonella species, Typhimurium and Enteritidis Multiplex PCR Assay with the SureTect Salmonella Infantis PCR Assay to simultaneously detect Salmonella spp. and identify the top 3 serotypes in poultry products linked to human illness: S. Typhimurium, S. Enteritidis, and S. Infantis.
The SureTect PCR System is an optimized solution combining speed and performance in an easy-to-use, cost-effective platform—giving you results you can trust.
Simple
- Easy-to-use and simple workflow
- Fast screening and identification of positive samples
- All-in-one workflow to get same-time results from a single test portion: Salmonella spp. and top 3 serotypes (S. Enteritidis, S. Typhimurium, and S. Infantis)
- Pre-dispensed and lyophilized reagents
Fast and Flexible
- Option to automate the lysis and PCR sample preparation
- Optimized workflow delivers on-screen results in as little as 18 hours
- Flexibility to run SureTect Salmonella species, Typhimurium and Enteritidis Multiplex PCR Assay with SureTect Salmonella Infantis Assay, simultaneously
- Test up to 96 samples in each run
- Choice of sealing lysis tubes with pierceable seals or caps
- Automatic lysis cycle
- Open PCR platforms
Smart
- Intuitive, user-friendly software delivers reliable results, with no subjective interpretation
- Actionable data supports fast decisions in a complex product supply chain
- IPC in every assay for complete confidence in results
The Thermo Scientific™ SureTect™ Salmonella Infantis PCR Assay Kit includes:
- Lysis Reagent 1 Tubes
- Proteinase K
- Salmonella Infantis PCR Tubes
- Thermo Scientific 96-Well Pierceable Seals
- PCR Caps
The SureTect Salmonella Infantis PCR Assay is validated on the Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR System with RapidFinder™ Analysis Software v3.0 or later or the Applied Biosystems™ 7500 Fast Food Safety 96-well Real-Time PCR instrument using Rapidfinder™ Express Software v2.0 or later, allowing you to test easily and confidently. After completing the PCR run, interpreted results are clearly displayed and can be reported, stored, printed, or downloaded as required.
Specifications
Specifications
| Content And Storage | Store at 2 to 8°C |
| Control Template | Internal Positive Control (IPC) |
| Detection Method | qPCR, RT-PCR |
| Form | Lyophilized |
| Format | Kit |
| PCR Method | qPCR |
| Technique | Real-time PCR |
| Disease | Salmonellosis |
| For Use With (Equipment) | Real-Time PCR (qPCR) System |
| High-throughput Compatibility | High-throughput Compatible, Automated Protocols |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.