Learn More
Description
Rapidly detect Salmonella species in food and environmental samples with the Thermo Scientific™ SureTect™ Salmonella species PCR Assay.
Detect Salmonella species in food and environmental samples with the Thermo Scientific™ SureTect™ Salmonella species PCR Assay which incorporates optimized primer and probe components for highly-specific detection of Salmonella species.
The assay's workflows have been validated for a broad range of human foods, pet food, including large test portions, and environmental samples by AOAC International and ISO 16140-2:2016 according to NF VALIDATION™ by AFNOR Certification and by MicroVal™ Certification. The assay is also certified for primary production samples by NF Validation™ by AFNOR Certification.
The new SureTect assay format incorporates color-coded plastics with orientation markings and breakable strip format and colored reagents and pierceable seals to enable an optimized workflow for maximum laboratory efficiency and productivity.
The SureTect System is an optimized solution combining speed and performance in an easy-to-use, cost-effective platform—giving you results you can trust.
Simple
- Easy-to-use with single enrichment step and pre-dispensed reagents
- Target-specific colored assay plastics
- Clearly oriented lysis and assay trays
- Color indicator in lysis reagent reducing possibility for human error
Fast and Flexible
- Option to automate the lysis and PCR sample preparation
- Optimized workflow delivers on-screen results in as little as 10 to 24 hours
- Maximum flexibility for operational efficiency
- Run all assays simultaneously in the same run
- Test up to 96 samples in each run
- Choice of pierceable seals or caps
- Detect multiple targets in the same run with universal PCR protocols across the range
Smart
- Intuitive, user-friendly software delivers reliable results, with no subjective interpretation
- Actionable data supports fast decisions in a complex product supply chain
- IPC in every assay for complete confidence in results
- NOW available with SureClean™ for your most challenging samples where free DNA can influence result performance
The Thermo Scientific SureTect Salmonella species PCR Assay Kit includes:
- Lysis Reagent 1 Tubes
- Proteinase K
- Salmonella species PCR Assay Tubes
- Thermo Scientific 96-Well Pierceable Seals
- PCR Caps
The SureTect Salmonella PCR Assay is validated on the Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR System using RapidFinder™ Analysis Software v3.0 or later or the Applied Biosystems™ 7500 Fast Food Safety Real-Time PCR Instrument using Rapidfinder™ Express Software v2.0 or later, allowing you to test easily and confidently. After completing the run, interpreted results are clearly displayed and can be reported, stored, printed, or downloaded as required.
The SureTect Salmonella PCR Assay has been tested extensively on a wide range of human food categories, including meat products, dairy products, fish and fishery products, fruits, and vegetables. The assay has also been tested on pet food and environmental samples, including primary products samples. Individual matrices tested in the method validation studies were selected based on their associated contamination by Salmonella and the known challenges linked with testing them by PCR methods. Individual matrices included but were not limited to ground beef, raw poultry, eggs, non-fat dried milk, and ready-to-eat meals.
Specifications
Specifications
| Product Type | SureTect Salmonella spp |
| For Use With (Application) | Food and Beverage |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.