Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SURF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501730189 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0189 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0189 Supplier Proteintech Group Inc Supplier No. 153791AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The SURF-1 protein demonstrates a vital role in the assembly of complex IV (CIV or COX) of the mitochondrial respiratory chain. Expressed in the inner mitochondrial membrane, mutations of the SURF-1 gene generally cause cytochrome c oxidase complex IV deficiency. Shortage of complex IV leads to Leigh syndrome, a severe neurological disorder. Leigh syndrome patients are usually subject to rapidly progressive encephalopathy, characterized by necrotic lesions in subcortical brain regions. SURF-1 mutations correlate to high post-implantation embryonic lethality as well as early-onset mortality of post-natal individuals. Considerable deficit in muscle strength and motor performance is also a profound and isolated defect of SURF-1 activity in skeletal muscle and liver. Heart, brain and skeletal muscle morphological abnormalities frequently occur due to SURF-1 mutations.

Specifications

Antigen SURF1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SURF1
Gene Accession No. P09925, Q15526, Q9QXU2
Gene Alias SURF 1, SURF1, surfeit 1, Surfeit locus protein 1
Gene Symbols Surf1
Host Species Rabbit
Immunogen SURF1 Fusion Protein Ag3959
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20930, 64463, 6834
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.