Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SURF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560677
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560677 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560677 Supplier Invitrogen™ Supplier No. PA560677
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSELPGDVRA FLREHPSLRL QTDARKVRCI LTGHELPCRL PELQVYTRGK KYQRLVRASP AFDYAEFEPH IVPSTKNPHQ LFCKLT Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 77%.

This gene shares a bidirectional promoter with surfeit 1 (SURF1; GeneID: 6834), which is located on the opposite strand. It encodes a conserved protein that is expressed in a variety of tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SURF2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SURF2
Gene Accession No. Q15527
Gene Alias SURF2; Surf-2; surfeit 2; surfeit gene 2; surfeit locus protein 2
Gene Symbols SURF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SURF2. Recombinant protein control fragment (Product #RP-93008).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6835
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.