Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SURF5 Antibody (PCRP-MED22-2A7), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314191H
This item is not returnable.
View return policy
Description
SURF5 Monoclonal antibody specifically detects SURF5 in Human samples. It is validated for ELISASpecifications
SURF5 | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
6837 | |
Store at 4°C in the dark. | |
Purified |
ELISA | |
PCRP-MED22-2A7 | |
ELISA | |
Med24, mediator complex subunit 22SURF5surfeit 5, mediator of RNA polymerase II transcription subunit 22, MGC48682, Surf-5, Surfeit locus protein 5 | |
Recombinant full-length human SURF5 protein (Uniprot: Q15528) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction